Would you like to log in again?
Tris HCl Buffer Solution, Quality Biological
Supplier: Quality Biological
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
351-006-131 351-006-721EA 351-092-101 351-091-101 351-048-101 351-006-721 351-007-101 351-006-101 351-050-101
10128-554EA 110.96 USD InStock
10128-554 10128-696 10128-498 10128-496 10128-396 10128-708 10128-394 10128-700 10128-698
Tris HCl Buffer Solution, Quality Biological
Buffers Biological Buffers
Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. These solutions of Tris hydrochloride provide convenient and highly purified solutions.
Tris is compatible with many enzymes in molecular biology, for example DNA modifying enzymes, because its buffering range is between 7.1 - 8.9. The lower pH 7.4 can be associated more with maintaining RNA, however, the pH 8.0 works equally as well with both DNA and RNA nucleic acids
Tris solutions are formulated into working concentrations for preparing the stacking and resolving portions of native or SDS-PAGE gels using discontinuous buffer systems according to Laemmli (1970) or Ornstein and Davis (1959).
Use Tris-HCl 1M pH 7.0-7.5 for DEAE Dextran-mediated transfection of non-adherent and adherent cells.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Ordering information: Please call a Avantor sales representative if you plan to order more than 100 bottles
Delivery information: Tris-HCl pH 7.0 1M, and Tris-HCl pH 8.8 1.5M require Special Production, 2-4 weeks for delivery
Caution: Research use only.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty