Would you like to log in again?
Dulbecco's Phosphate Buffered Saline (DPBS), Quality Biological
Supplier: Quality Biological
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
114-057-131CS 114-057-151 114-154-491 114-154-151 119-068-131 119-068-151 114-057-491 119-068-491 114-057-101 114-154-131 114-057-131 114-057-101CS 114-059-101 119-068-101
10128-840EA 54.27 USD InStock
10128-840 10128-750 10128-718 76716-354 10128-716 10128-738 10128-748 10128-846 10128-714 10128-746 10128-844 10128-722 10128-842 10128-720
Dulbecco's Phosphate Buffered Saline (DPBS), Quality Biological
Buffers Cell Culture Buffers
DPBS is a balanced salt solution (BSS) used for the handling and culturing of mammalian cells. Calcium and magnesium facilitate cell binding and clumping. DPBS without these ions can be used to wash and rinse suspended cells.
  • Without phenol red
DPBS can be used to provide a buffer system to maintain cell culture media in the physiological pH range of 7.0 to 7.6
DPBS serves as an irrigating, transporting or diluting fluid for mammalian cells, while maintaining cell tonicity and viability for a limited period of time.
A 1X working solution is recommended for application usage.
DPBS is also used as a diluent of fresh red blood cells to study the trapping of red blood cell aggregate, to wash the wells of microtiter plates, and to wash cells during preparation and serial transfer.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research use only.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty