Would you like to log in again?
Phosphate Buffered Saline (PBS), Quality Biological
Supplier: Quality Biological
114-058-491 119-069-101CS 114-056-491 119-069-161 114-056-151 119-069-151 114-056-101CS 114-058-101 114-058-151 114-056-101 119-069-131CS 114-058-131CS 114-058-131 114-058-101CS 119-069-101 119-069-491 114-058-161 119-069-131
10128-730CS 570.11 USD InStock
10128-730 10128-850 10128-674 10128-860 10128-728 10128-848 10128-858 10128-726 10128-736 10128-856 10128-724 10128-712 10128-734 10128-854 10128-710 10128-732 10128-676 10128-852
Phosphate Buffered Saline (PBS), Quality Biological
Buffers Cell Culture Buffers
PBS (phosphate buffered saline) is a balanced salt solution used for a variety biological and cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.
  • Endotoxin and Performance tested
PBS is also useful in immunological applications, such as Western Blotting.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty