Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trypsin (0.05%), EDTA (0.1%)

Supplier: Quality Biological
118-087-721 118-087-721EA
10128-784PK 92.55 USD InStock
10128-784 10128-786
Trypsin (0.05%), EDTA (0.1%)
Enzymes
Trypsin is a proteolytic enzyme used to detach adherent cell from culture vessel surfaces. Typical use includes removing adherent cells from a culture surface. The concentration of trypsin necessary to dislodge cells from their substrate is dependent primarily on the cell type and the age of the culture.

  • With EDTA
  • With Phenol Red

Trypsin is a serine protease derived from porcinepancreas. It is a single chain polypeptide of 223 amino acid residue with substrate specificity based on positively charged Lysine and Arginine side chains. Trypsin predominantly cleaves peptide chains at the carboxyl sides of Lysine and Arginine, except when either is followed by Proline. It is most commonly used for dissociation and disaggregation of adherent cells.

Ethylenediaminetetraacetic acid (EDTA), a chelating agent often added to enhance enzymatic activity of trypsin solution. EDTA acts by neutralizing calcium and magnesium ions that enhance cell-to-cell adhesion. Polyvinylpyrrolidone (PVP) a synthetic polymer is known to increase cell viability especially in low serum or serum free condition.

Trypsin 0.05% – EDTA 0.1%, assists in detaching normal and transformed cells from vessel surfaces.

Sterile Filtered
Harmonization Code: 3822.00.5090
ECCN: EAR99

Certifications: Manufactured in an ISO 9001:2015 certified facility.

Ordering information: *NOTE: During shipping, the trypsin might change color from pink to yellow due to exposure to carbon dioxide released from the dry ice in the shipping container. In our experience, this discoloration does not effect the functionality of the trypsin.

Caution: Research Use Only

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty