Would you like to log in again?
ACK Lysing Buffer, Quality Biological
Supplier: Quality Biological
118-156-101CS 118-156-721 118-156-101 118-156-721EA
10128-808EA 69.73 USD InStock
10128-808 10128-806 10128-804 10128-802
ACK Lysing Buffer, Quality Biological
Buffers Extraction Buffers
Ammonium-Chloride-Potassium (ACK) Lysing Buffer is used for the lysis of red blood cells in samples containing white blood cells, such as EDTA-treated whole blood, buffy coats, and bone marrow.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty