Would you like to log in again?
Thermo Pipette Calibration Technical Services, Thermo Scientific
Supplier: Thermo Scientific
VFC-2X5-DMC GXP-3X5-DMC VFC-2X10-DSC PMC-2X5-DMC GXP-3X10-DMC PMC-2X5-DSC GXP-3X10-DSC VFC-2X10-DMC GXP-3X5-DSC VFC-2X5-DSC PMC-2X10-DMC PMC-2X10-DSC
10129-286EA 77.32 USD InStock
10129-286 10129-288 10129-294 10129-282 10129-284 10129-448 10129-292 10129-312 10129-466 10129-314 10129-468 10129-446
Thermo Pipette Calibration Technical Services, Thermo Scientific
Services
Whether it's cGLP, cGMP, critical research, academia, or special requirements, our experienced service personnel are highly trained in providing exceptional care for Pipettes
For fast, convenient Pipette service, order a free return kit to be mailed to you. Kits include: Pipette Service Return Form, Pipette Service Decontamination Form, Insulated foam box to protect Pipettes during shipment, and a Prepaid Airway Bill. There is no charge to you for the shipping.
Three levels of service for Pipettes are offered. The Basic Validation-Fast Check Plan combines a thorough external Pipette inspection with gravimetric validation to determine if desired accuracy and precision is achieved. This plan includes a validation pass/fail report. This offering is designed for customers that need: an interim validation check that occurs in-between routine scheduled preventive maintenance and calibration; a dropped, disassembled, or standard performance update; or a cost-friendly service option. The Basic Plan is quick, convenient, and effective.
The Preventive Maintenance and Calibration Plan provides a thorough inspection, preventive maintenance, replacement of O-Rings as needed, and calibration. This plan includes a calibration certificate and sticker. This offering is ideal for customers that: do not require as-received data; require Pipette inspection, liquid leak test, routine preventive maintenance and replacement of O-Rings (as needed); calibration; checked routinely: Pipette body and display, tip ejection and ejector mechanism, tip cone(s), piston(s) assembly, O-Ring(s), and springs; checked as applicable: plunger bottom, plunger rod, volume lock, shaft coupling nut, seal(s), lubrication, battery recharging, electronics functionality, and motor movement. The Preferred Plan is thorough, effective, and dependable.
The GxP Preventive Maintenance and Calibration Plan provides a comprehensive service which includes gravimetric evaluation of Pipette performance prior to any inspection, preventive maintenance, or replacement of O-Rings as needed. This plan includes a calibration certificate and sticker. This offering is designed for customers that: require as-received data; require Pipette inspection, liquid leak test, routine preventive maintenance, and replacement of O-Rings (as needed); calibration; work in a regulated environment where Pipette accuracy and precision are critical; need to minimize costly OOTs (Out of Tolerance) that could invalidate research; checked routinely: Pipette body and display, tip ejection and ejector mechanism, tip cone(s), piston(s) assembly, o-ring(s), and springs; checked as applicable: plunger bottom, plunger rod, volume lock, shaft coupling nut, seal(s), lubrication, battery recharging, electronics functionality, and motor movement. The Premium Plan is demanding, thorough, and reliable.
For over twenty-five years, we offer quality-minded service that meets your diverse needs. As a result, you have the assurance that your Pipettes have the accuracy you need when ready for use.
Ordering information: Calibration services are only available in the United States. Pipettes should be in working order prior to sending for calibration.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty