Would you like to log in again?
Reusable Very High Pressure (VHP) Fittings, IDEX Health and Science
Supplier: IDEX Health & Science
Upchurch Scientific®
VHP-3200
10129-530EA 72.24 USD InStock
10129-530
Reusable Very High Pressure (VHP) Fittings, IDEX Health and Science
Fittings and Connectors
IDEX Health and Science very high pressure (VHP) fittings are designed to withstand the extreme operating pressures of today's modern UHPLC systems
These Reusable VHP fittings can be reused when following the tightening torque specifications listed for each product. With a polymer front ferrule, there is no damage to the tubing or the receiving port, adding to the usable lifetime of these products. These fittings will allow for quick column, sample loop, inline filter or tubing changes with minimal downtime.
This patent-pending line of ground-breaking fittings provides reliable connections with stainless steel (and stainless steel jacketed) tubing, facilitating reusable fluidic pathway connections where system pressures can reach 10,000 psi or more.
Torque: 3.5 in-lbs (0.4 N-m) Hand-Tight / 10 in-lbs (1.10 N-m) Tool Tight
Certifications: Complies with Directive 2011/65/EU, the Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment (RoHS) of the European Union. Complies with Regulation 1907/2006 - Registration, Evaluation, Authorization and the Restriction of Chemicals (REACH) of the European Union.
Ordering information: Part numbers reference individual fittings. Tubing is not included with these fittings and should be ordered separately. Any wrenches or similar tools that may be needed for complete tightening also must be purchased separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty