Would you like to log in again?
NUT PEEK BLK METRC PK10
Supplier: IDEX HEALTH & SCIENCE LLC
Black PEEK Flangeless fitting, Nut, M6x1 threaded, PEEK, BLACK, 0. 125 in, Pack of 10^
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
NUT PEEK BLK METRC PK10 10129-534 P-347X Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty