Would you like to log in again?
Dewax and HIER H Buffer, Epredia
Supplier: Epredia
TA-999-DHBH
10129-598CS 953.68 USD InStock
10129-598
Dewax and HIER H Buffer, Epredia
Stains Microscopy and staining reagents
Epredia™ Dewax and Heat Induced Epitope Retrieval (HIER) H Buffer simultaneously automate deparaffinization, rehydration and epitope retrieval in the Epredia™ PT Module.
Automated deparaffinization (dewaxing) and rehydration of IHC slides is an ideal way to decrease turn-around time and improve lean workflow. Aqueous deparaffinization is the method of choice for IHC instruments today but often yields incomplete deparaffinization.
The Dewax and Heat Induced Epitope Retrieval (HIER) Buffer simultaneously automate deparaffinization, rehydration and epitope retrieval in the Epredia PT Module. The new proprietary formulation eliminates manual steps and improves turnaround time. In addition the results are concordant to traditional methods meaning they can be quickly integrated into routine workflow.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty