Would you like to log in again?
SUPRA-CLEAN SPE COLUMNS C18-S 500MG/3ML
Supplier: PERKINELMER U.S. LLC
Column^ SPE Supra-Clean* C18-S, 500 mg/3 mL, Spherical Silica-based Solid Phase Extraction (SPE) Columns are Can be useded for extracting mid-polar to non-polar compounds when analyzing drugs and drug metabolites in biological matrices.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SUPRA-CLEAN SPE COLUMNS C18-S 500MG/3ML 10131-450 N9306438 Pack of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty