Would you like to log in again?
Brain Heart Infusion (BHI) Broth, Dry Media, Teknova
Supplier: Teknova
B9505 B9500 B9505 B9500
101324-842EA 353.08 USD InStock
101324-842 CA101324-842 CA101319-540 101319-540
Brain Heart Infusion (BHI) Broth, Dry Media, Teknova
Media for Microbiology Dehydrated Media for Microbiology
Brain Heart Infusion Broth is a general-purpose medium suitable for the cultivation of a wide variety of organisms, including bacteria, yeasts and molds.
  • Cultivates a wide variety of organisms
  • Made of 3.7% Brain Heart Infusion Powder
Ambient shipping conditions. Store at room temperature.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty