Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloramphenicol 34 mg/mL in ethanol 100%

Supplier: TEKNOVA INC
Synonyms: Chloromycetin®(R,R)-ChloramphenicolTega-Cetin

C0322
101319-572EA 52.31 USD InStock
101319-572
Chloramphenicol 34 mg/mL in ethanol 100%
Chloramphenicol
Chloramphenicol is a broad-spectrum antibiotic effective against most strains of Gram-positive and Gram-negative bacteria. It is bacteriostatic but may be bactericidal at high concentrations and is lipid-soluble, allowing it to diffuse through the bacterial cell membrane.


Formula: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MW: 323.13 g/mol
MDL Number: MFCD00078159
CAS Number: 56-75-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty