Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Imidazole 2M for biochemistry, sterile Imidazole 2M pH 7.5, Sterile

Supplier: TEKNOVA INC
Synonyms: Glyoxaline1H-ImidazoleGlyoxalamine

I6902
101319-916EA 516.56 USD InStock
101319-916
Imidazole 2M for biochemistry, sterile Imidazole 2M pH 7.5, Sterile
Imidazole


Formula: C₃H₄N₂
MW: 68.08 g/mol
MDL Number: MFCD00005183
CAS Number: 288-32-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty