Would you like to log in again?
Mueller Hinton II Cation Adjusted Broth, Teknova
Supplier: Teknova
M5887 M5887
101320-364EA 496.92 USD InStock
101320-364 CA101320-364
Mueller Hinton II Cation Adjusted Broth, Teknova
Media for Microbiology Prepared Media for Microbiology
Mueller Hinton II cation adjusted broth contain 1.75% acid hydrolysis of casein, 0.3% beef extract, 0.15% starch, cation-adjusted with calcium and magnesium as per NCCLS standard M7.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty