Would you like to log in again?
Reaction Flask Clamp, DWK Life Sciences
Supplier: DWK Life Sciences
613750-0000
10135-804CS 356.88 USD InStock
10135-804
Reaction Flask Clamp, DWK Life Sciences
Clamps General Purpose Clamps
Aluminum construction to connect tops to flasks.
Wing nuts or thumb screws are spaced to assure uniform compression for sealing the system.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty