Would you like to log in again?
EE Broth Mossel, USP, Hardy Diagnostics
Supplier: Hardy Diagnostics
U291
10135-884PK 248.06 USD InStock
10135-884
EE Broth Mossel, USP, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For use in the selective enrichment and detection of Enterobacteriaceae, especially in foods.
For use in the selective enrichment and detection of Enterobacteriaceae, especially in foods.
Ordering information: Minimum order of 12 packs
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty