Would you like to log in again?
Letheen Broth Modified, Hardy Diagnostics
Supplier: Hardy Diagnostics
U309 U293 U397
76203-718PK 198.08 USD InStock
76203-718 10135-888 76299-206
Letheen Broth Modified, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Used as a diluent in the preparation of samples for microbial analysis.
  • A crystal clear glass or polycarbonate construction provides complete visibility
  • Features a streamlined design with no handle, making it easier to store and clean
Letheen Broth, Modified is for use in microbial recovery from personal care products and for the neutralization of quaternary ammonium compounds in the testing of germicidal activity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty