Would you like to log in again?
ELISA Assays for Infectious Disease and Autoimmune, Gold Standard Diagnostics
Supplier: Gold Standard Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
EIA-1292 9165 GSD01-3003 720-430 720-450 720-700 GSD01-120 GSD01-100 720-250 720-240 GSD01-460 720-460 GSD01-1001 ORG522A 9177 GSD01-160 GSD01-3000 720-350 9166 720-290 GSD01-140 FCCP600 GSD01-150 720-270 GSD01-470 9178 GSD01-1000 720-280 EL0920G GSD01-110 ORG522G 720-230 EIA-3954 EL0910G 720-260 GSD01-130 GSD01-200 GSD01-3002 GSD01-490 ORG522M GSD01-180 GSD01-480
10066-194KT 592.46 USD InStock
10066-194 10066-196 10066-230 10066-198 10066-232 10066-178 10066-212 10066-214 10135-958 10135-956 10135-918 10066-218 10135-938 10111-850 10135-920 10119-432 10119-434 10119-436 10066-190 10111-990 10119-438 10066-192 10066-188 10119-430 10066-200 10119-452 10066-222 10066-224 10135-946 10135-924 10066-202 10135-922 10135-928 10135-926 10066-208 10135-954 10135-930 10119-422 10119-424 10119-426 10066-180 10119-428
ELISA Assays for Infectious Disease and Autoimmune, Gold Standard Diagnostics
Assays ELISAs
Gold Standard Diagnostics offers a comprehensive menu of ELISA assays for Infectious Disease & Autoimmune diagnostic testing
Most kits feature interchangeable reagent components, consistent protocols, break-away wells.
Our comprehensive menu of assays fits the needs of virtually any laboratory.
Certifications: FDA Cleared or Exempt.
Ordering information: Package contains Microtiter Plate(s), Dilution Buffer, Wash Buffer, Controls, Conjuage and Stop Solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty