Would you like to log in again?
Amp'd® ELISA Signal Amplification Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT-100-0005 KIT-100-0001
10135-982EA 200 USD InStock
10135-982 10135-984
Amp'd® ELISA Signal Amplification Kit, Enzo Life Sciences
Assays ELISAs
Amp'd® ELISA Signal Amplification Kit provides up to 50-fold increase in sensitivity over traditional ELISAs while detecting lower concentrations of target in samples.
For the amplification of ELISA sensitivity designed to replace traditional alkaline phosphatase (AP) substrates.
The Amp'd® ELISA Signal Amplification kit is designed to replace traditional alkaline phosphatase (AP) substrates, such as pNPP (p-Nitrophenyl phosphate), with a combination substrate and amplifier system that results in greater sensitivity when compared to a classic substrate enzyme linked-immunosorbent assay (ELISA).
Ordering information: Contains Signal Amplification Substrate, Amplifier, Substrate Diluent, and Amplifier Diluent.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty