Would you like to log in again?
Amp'd® HSP70 High Sensitivity ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT-101-0001
10135-994EA 849 USD InStock
10135-994
Amp'd® HSP70 High Sensitivity ELISA Kit, Enzo Life Sciences
Assays ELISAs
Ultra-sensitive (7 pg/ml)Amp'd® HSP70 high sensitivity ELISA kit enabling the ability to use less sample and detect both baseline and upregulated levels of human, mouse and rat Hsp70 (Hsp72), a major chaperone, cancer biomarker, and key cell stress regulator.
For the quantitative determination of normal and upregulated (stressed) levels of HSP70 in serum and plasma of human, mouse, and rat origin.
The Amp'd® HSP70 high sensitivity ELISA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4.5 hours.
Ordering information: Contains Microtiter Plate, Assay Buffer 28, Standard, Wash Buffer Concentrate, Antibody, Conjugate, and Amp'd (Signal Amplification) Substrate, Amplifier, Substrate Diluent, Amplifier Diluent, and Stop Solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty