Would you like to log in again?
Organelle-ID RGB® Reagent IV for Microscopy, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-53009-C200
10136-044EA 468.29 USD InStock
10136-044
Organelle-ID RGB® Reagent IV for Microscopy, Enzo Life Sciences
Protein Linking and Labeling Systems
Mixture of fluorescent dyes for detection of lysosomes, endoplasmic reticulum and nucleic acids
The Organelle-ID RGB® Reagent IV is a mixture of cell-permeable red fluorescent endoplasmic reticulum dye, green fluorescent lysosomal dye and blue fluorescent nucleic acid dye. The staining pattern arising from the combination of these three dyes permits visualization of the target organelles by fluorescence/confocal microscopy. The reagent, supplied as a 500X solution, is sufficient for 1000 microscopy assays. The single-tube format makes this multi-organelle stain reagent easy to use.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty