Would you like to log in again?
VWR® Tube Rotator and Rotisseries
Supplier: VWR
VWR®
H5600A-VWR
10136-084EA 662.72 USD InStock
10136-084
VWR® Tube Rotator and Rotisseries
Shakers and Mixers Rotating/Rocking Shakers
The unique design of the VWR® Tube Rotator allows for a variety of tube sizes and mixing applications to be performed efficiently on one instrument.
  • Ideal for biochemistry, molecular biology, histochemistry, and clinical applications
  • Unique adjustable rotisseries allow for horizontal or vertical tube mixing
  • Safe for coldroom or incubator use
Rotisserie plates are attached to a fixed center spindle and can be mixed and matched
Placing the rotisserie plate perpendicular to the center spindle produces a simple tube rotation that is gentle enough for hybridization applications. Turning the plate parallel to the center spindle produces a more vigorous, end-over-end tumbling motion that is often required for mixing blood samples or protein precipitation. Plates can be adjusted to any position between these extremes to produce a variety of mixing levels, with no tools required.
The tube rotator is a compact, portable instrument that can be easily transported from bench to coldroom, incubator, or biological hood. The molded housing and rotisseries are designed for simple cleaning and decontamination.
The two halves of the rotisserie can be adjusted independently of one another to allow for simultaneous horizontal and vertical mixing. Rotisserie speed is fixed at 18rpm (20rpm for 230V model).
Ordering information: All tube rotators are supplied with a rotisserie assembly that holds thirty-six 1.5/2.0 mL tubes as well as 2 extra paddles for 3 x 50 mL tubes and a combination of 5 x 15 mL tubes or 8 x 5/7 mL tubes as shown. Additional rotisserie assemblies are sold separately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty