Would you like to log in again?
Sampling Probes Assemblies for Autosamplers, PerkinElmer
Supplier: Perkin Elmer
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
B3000160 B3001771 B3001772 N8145139 B3001769 B3001770 N0777434 B3000161 B3000152 B3000159
97013-200EA 772.77 USD InStock
97013-200 10136-176 10136-420 97013-190 97013-208 97013-192 10137-644 97013-182 97013-184 10136-728
Sampling Probes Assemblies for Autosamplers, PerkinElmer
Electrodes Other Electrodes
Sampling probe assemblies for AS-90/90A/90plus/91/93plus/S10 autosampler.
Corrosion-resistant probes are suitable for inorganic acids and most organic solvents, except NMP.
PTFE probes provide laminar flow and are the most resistant to chemical degradation.
Delivery information: Corrosion-resistant sampling probe assemblies includes sampling Probe with FEP tube, screw fitting and capillary tubing assembly.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty