Would you like to log in again?
Fluid A, Bottles, Northeast Lab Services
Supplier: Northeast Lab Services
B8110
101373-778EA 8.46 USD InStock
101373-778
Fluid A, Bottles, Northeast Lab Services
Media for Microbiology Prepared Media for Microbiology
Fluid A includes a serum bottle, a rubber stopper, and an aluminum crimp cap.
  • Rubber Stopper
  • Aluminum Crimp Cap
  • Fill Volume: 100 mL
The serum bottle is 4 oz., and Fluid A has a fill volume of 100 mL.
NEL manufactures over 500 proprietary products and over 1300 classic media formulations for clients in the pharmaceutical, hospital, food and beverage, cosmetic, and educational research industries.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty