Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Modified Oxford Medium, Plates, Northeast Lab Services

Supplier: Northeast Lab Services
P1452
101374-034PK 168.86 USD InStock
101374-034
Modified Oxford Medium, Plates, Northeast Lab Services
Media for Microbiology Prepared Media for Microbiology
Modified Oxford Medium is contained in a standard Petri Dish.

  • Standard Petri Dish
  • Fill Volume: 20 mL
  • Prepared Medium
  • Size: 15 mm x 100 mm

The petri dish is 15 mm x 100 mm, and the medium's fill volume is 20 mL.

NEL's plated media operation utilizes carefully selected and validated ingredients for contaminant-free products with a high degree of consistency from batch to batch.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty