Would you like to log in again?
Nutrient Agar, Plates, Northeast Lab Services
Supplier: Northeast Lab Services
P1491
101374-050PK 117 USD InStock
101374-050
Nutrient Agar, Plates, Northeast Lab Services
Media for Microbiology Prepared Media for Microbiology
Nutrient Agar is contained in a standard Petri Dish.
  • Standard Petri Dish
  • Fill Volume: 24 mL or 20 mL
  • Prepared Medium
  • Various Sizes Available
NEL's plated media operation utilizes carefully selected and validated ingredients for contaminant-free products with a high degree of consistency from batch to batch.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty