Would you like to log in again?
Anaerobic Blood Agar, Plates, Northeast Lab Services
Supplier: Northeast Lab Services
P5010
101374-320PK 245.45 USD InStock
101374-320
Anaerobic Blood Agar, Plates, Northeast Lab Services
Media for Microbiology Prepared Media for Microbiology
For the cultivation of anaerobes in clinical specimens.
  • Enriched, non-selective medium
  • Isolation of obligatory anaerobic organisms
An enriched, non-selective medium for isolation of a wide variety of obligatory anaerobic organisms, especially ones found in clinical specimens.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty