Would you like to log in again?
Simmons Citrate Slant, Tubes, Northeast Lab Services
Supplier: Northeast Lab Services
T1730
101374-800PK 119.43 USD InStock
101374-800
Simmons Citrate Slant, Tubes, Northeast Lab Services
Media for Microbiology Prepared Media for Microbiology
Simmons tubes have a sturdy screw cap.
  • Screw Caps
  • Tubed Agar Serves a Variety of Lab Functions
  • Glass Tubes
  • Fill Volume: 3 mL
  • Size: 13 mm x 100 mm
From simple fermentation tubes to highly specialized clinical diagnostic media, NEL produces media tubes for a wide variety of clients including private laboratories, food processors, clinics and hospitals, academic institutions, and government agencies.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty