Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium dodecyl sulfate 98+% (by acidimetry)

Supplier: Thermo Fisher Scientific
Warning
Synonyms: Lauryl sulfate lithium saltLithium lauryl sulphateDodecyl sulfate lithium saltLithium lauryl sulfateDodecyl sulphate lithium saltLauryl sulphate lithium saltDodecyl sulphuric acid lithium saltDodecyl sulfuric acid lithium salt

J32816-A1 J32816-14 J32816-22
101375-330EA 277.85 USD InStock
101375-330 101174-788 103728-124
Lithium dodecyl sulfate 98+% (by acidimetry)
Lithium dodecyl sulphate


Formula: C₁₂H₂₅LiO₄S
MW: 272.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007467
CAS Number: 2044-56-6
EINECS: 218-058-2
UN: 1325
ADR: "4.1",III
Order Now
Specification Test Results
Form White crystalline powder
Assay (unspecified) ≥98% (acidity)
Assay (unspecified) Absorbance (230 nm, 3%, 1 cm) ≤ 0.2
Assay (unspecified) Absorbance (280 nm, 3%, 1 cm) ≤ 0.1
Assay (GC) (C12) ≥98%
Chloride content ≤ 0.01%
Loss on Drying ≤ 1%
Trace Metal: Copper (Cu) ≤ 5 ppm
Trace Metal: Iron (Fe) ≤ 1 ppm
Trace Metal: Lead (Pb) ≤ 5 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty