Would you like to log in again?
TUBE COMBUSTION QUARTZ F/CHN ANALYSIS
Supplier: PERKINELMER U.S. LLC
Combustion Tube for CHN Analysis, Quartz^ Combustion tubes are composed of pure fused quartz which is 99% Si02. Fused quartz has been chosen because of its very high purity and low thermal expansion rate, Package 10
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBE COMBUSTION QUARTZ F/CHN ANALYSIS 10138-298 N2410153 Pack of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty