Would you like to log in again?
QUARTZ 10 MM BENDING PLATFORM FOR DMA 7
Supplier: PERKINELMER U.S. LLC
Bending Platform^ Quartz 10 mm Bending Platform for DMA 7/DMA 7e 3-Point Measuring System and TMA 7. This tube differs from the standard sample tube in that the 10mm is permanently fused to the base of the sample tube
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
QUARTZ 10 MM BENDING PLATFORM FOR DMA 7 10138-670 N5391181 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty