Would you like to log in again?
MicroFlow-ST Concentric Nebulizer for PerkinElmer ICP-MS, PerkinElmer
Supplier: PerkinElmer
N8122192
10138-934EA 3664.33 USD InStock
10138-934
MicroFlow-ST Concentric Nebulizer for PerkinElmer ICP-MS, PerkinElmer
Atomizers
This PFA-ST high efficiency microFlow nebulizer is for use with all ELAN® and NexION® ICP-MS series instruments. The PFA-ST nebulizer features high purity, HF-resistance and high performance, along with an exchangeable external sample uptake capillary. The sample uptake rate is controlled by the dimension of the external capillary or probe. It is chemically resistant and ideal for strong acids, alkalis, and organics.
  • Chemically resistant: Ideal for strong acids, alkalis, organics
  • Resistant to clogging: Reliably self-aspirated or pumped
  • HF resistant
MicroFlow nebulizers are constructed entirely from chemically resistant fluoropolymers, ideal for strong acids, alkalis, organics, high salt solutions and high solid concentrations. They are resistant to clogging, are reliably self-aspirated or pumped and produce a fine aerosol for high transport efficiency and high sensitivity.
This nebulizer is included in all PC3 sample introduction kits and in the PFA-sapphire HF-resistant sample introduction kit.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty