Would you like to log in again?
Ilford Multigrade IV RC Deluxe Photographic Papers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
83021-10 83020-10 83020-25 83021-1M
101381-622PK 1522.44 USD InStock
101381-622 101381-620 101381-618 101381-616
Ilford Multigrade IV RC Deluxe Photographic Papers, Electron Microscopy Sciences
Film Processing Supplies
This is a direct replacement for the Kodak Polycontrast IV RC B&W Papers. Neutral tone – Variable contrast. MULTIGRADE IV RC Deluxe has the standard weight of (190g/m2)with a resin coated base.
  • Resin Coated – Black & White
  • Non-Developer Incorporated
  • 12 Contrast Grades with MULTIGRADE Fibers
  • Glossy Surface
  • Compatible with the Kodak OC Filter
  • Dish/Tray Processing
  • Developer: Mutigrade, Bromophen or PQ Universal
  • Fixer: Ilford Rapid Fixer or Hypam
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty