Would you like to log in again?
Autosampler Vials, PFA, PerkinElmer
Supplier: PerkinElmer
N0777401
10139-362PK 308.74 USD InStock
10139-362
Autosampler Vials, PFA, PerkinElmer
Vials
PerkinElmer produces a wide range of PFA autosampler vials in a variety of shapes and sizes, to cover virtually every application.
  • Preferred for semiconductor use
  • Translucent siding for optimal visibility
  • Conical interior
PFA vials are preferred for use with semiconductors, and PFA alternatives exist for a majority of polypropylene vials.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty