Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD5 Mouse Antibody (APC (Allophycocyanin)) [clone: UCHT2]

Supplier: Tonbo Biosciences
20-0059-T025 20-0059-T100
10140-994EA 83.4 USD InStock
10140-994 10140-996
Anti-CD5 Mouse Antibody (APC (Allophycocyanin)) [clone: UCHT2]
Antibodies
The UCHT2 antibody is specific for human CD5, a 67 kD transmembrane glycoprotein, that is expressed on most thymocytes, mature T cells, and a subset of B cells. CD5 is involved in modulating antigen receptor signaling in both T and B cells and binds to CD72, expressed on B cells.

Type: Primary
Antigen: CD5
Clonality:
Clone: UCHT2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty