Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD5 Mouse Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: UCHT2]

Supplier: Tonbo Biosciences
65-0059-T025 65-0059-T100
10141-068EA 233 USD InStock
10141-068 10141-066
Anti-CD5 Mouse Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: UCHT2]
Antibodies
The UCHT2 antibody is specific for human CD5, a 67 kD transmembrane glycoprotein, that is expressed on most thymocytes, mature T cells, and a subset of B cells. CD5 is involved in modulating antigen receptor signaling in both T and B cells and binds to CD72, expressed on B cells.

Type:
Antigen: CD5
Clonality:
Clone: UCHT2
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty