Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PTPRC Rat Antibody (redFluor® 710) [clone: 30-F11]

Supplier: Tonbo Biosciences
80-0451-U025 80-0451-U100
10141-104EA 147 USD InStock
10141-104 10141-102
Anti-PTPRC Rat Antibody (redFluor® 710) [clone: 30-F11]
Antibodies
The 30-F11 antibody reacts with mouse CD45, which is one of the most abundant hematopoietic markers and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). CD45 is a protein tyrosine phosphatase existing in several isoforms, each being generated and expressed in cell-specific patterns. With its broad cell distribution, CD45 is critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RA, CD45RB and CD45RO.

The 30-F11 antibody is widely used as a leukocyte marker for B cells, T cell subsets and NK cell subsets.

Type: Primary
Antigen: PTPRC
Clonality:
Clone: 30-F11
Conjugation: redFluor® 710
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty