Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rat IgG2b Isotype Control [clone: LTF-2] (redFluor® 710)

Supplier: Tonbo Biosciences
80-4031-U100
10141-118EA 114 USD InStock
10141-118
Rat IgG2b Isotype Control [clone: LTF-2] (redFluor® 710)
Antibodies
The LTF-2 immunoglobulin is useful as an isotype-matched control. The LTF-2 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2b antibodies.

Type: Control
Antigen: LTF-2
Clonality:
Clone: LTF-2
Conjugation: redFluor® 710
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty