Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MHC Class II (I-A/I-E) (M5/114.15.2) Rat Antibody [clone: M5/114.15.2] (redFluor™ 710)

Supplier: Tonbo Biosciences
80-5321-U025 80-5321-U100
10141-120EA 151 USD InStock
10141-120 10141-260
Anti-MHC Class II (I-A/I-E) (M5/114.15.2) Rat Antibody [clone: M5/114.15.2] (redFluor™ 710)
Antibodies
The M5/114.15.2 antibody reacts with mouse MHC Class II alloantigens I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek, as well as being cross-reactive with mouse cells of H-2p and H-2r haplotype. MHC Class II is widely expressed by mouse immune cells bearing these alloantigens, including T and B cells, monocytes, macrophages and dendritic cells. The antibody does not react with the following alloantigens: I-Af, I-Ak, I-As, or NOD H-2g7.

The M5/114.15.2 antibody may be used for analysis of mouse cells expressing MHC Class II alloantigens as described. Please note that the M5/114.15.2 clone may also be referred to as M5/114 in the literature.

Type: Primary
Antigen: MHC Class II (I-A/I-E)
Clonality:
Clone: M5/114.15.2
Conjugation: redFluor™ 710
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty