Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD2 Rat Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RM2-5]

Supplier: Tonbo Biosciences
35-0021-U500 35-0021-U100 35-0021-U025
10141-148EA 212 USD InStock
10141-148 10141-146 10141-144
Anti-CD2 Rat Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RM2-5]
Antibodies
The RM2-5 antibody reacts with mouse CD2, an approximately 50 kDa glycoprotein, and a member of the Ig superfamily. CD2, also known as LFA-2, is a receptor for CD48 in the mouse and is expressed on the cell surface of all mouse lymphocytes. CD2 mediates adhesion between cells and is involved in T cell activation.

RM2-5 is reported to block CD2-mediated cell:cell adhesion. Please choose the appropriate format for each application.

Type:
Antigen: CD2
Clonality: 0
Clone: RM2-5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty