Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD40L Armenian Hamster Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MR1]

Supplier: Tonbo Biosciences
35-1541-U100 35-1541-U025
10141-156EA 114 USD InStock
10141-156 10141-154
Anti-CD40L Armenian Hamster Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MR1]
Antibodies
The MR1 antibody is specific for mouse CD154, a 39 kD glycoprotein that is primarily expressed on activated T cells. CD154 (CD40 ligand, gp39) binds to CD40 expressed on antigen presenting cells and mediates important costimulatory functions between these cells. Signals generated via cognate interactions between CD154-CD40 on activated T and B cells are required for T-dependent B cell responses. The MR1 antibody is reported to block interaction of CD154 with CD40.

Type:
Antigen: CD40L
Clonality:
Clone: MR1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty