Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FASLG Mouse Antibody [clone: NOK-1]

Supplier: Tonbo Biosciences
40-9919-U100
10141-188EA 125 USD InStock
10141-188
Anti-FASLG Mouse Antibody [clone: NOK-1]
Antibodies
The NOK-1 antibody reacts with human CD178 (Fas ligand) in both membrane bound and soluble forms. Fas ligand is a 40 kDa transmembrane glycoprotein, a member of the TNF family, and is expressed by activated T and NK cells, neutrophils, and monocytes. Interactions between CD178 (Fas ligand) and CD95 (Fas) induce a program of apoptosis and play a key role in immune regulation and homeostasis. The extracellular domain of human CD178 can be cleaved from the surface by matrix metalloproteinases (MMPs) resulting in a 26 kDa soluble protein.

Type: Primary
Antigen: CD178 (Fas ligand)
Clonality:
Clone: NOK-1
Conjugation: In Vivo Ready™
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty