Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CSF1R Rat Antibody (PE (Phycoerythrin)) [clone: AFS98]

Supplier: Tonbo Biosciences
50-1152-U100 50-1152-U025
10141-202EA 70.2 USD InStock
10141-202 10141-204
Anti-CSF1R Rat Antibody (PE (Phycoerythrin)) [clone: AFS98]
Antibodies
The AFS98 antibody is specific for mouse CD115, also known as Colony-Stimulating Factor-1 Receptor (CSF-1R), a 145 kDa receptor from the PDGF receptor family. Receptor activation by the ligands IL-34 or CSF-1 (M-CSF) occurs via homodimerization of CD115 and subsequent tyrosine phosphorylation and ubiquitination of intracellular domains. CD115 signaling promotes differentiation of myeloid precursors, as well as the continued regulation of proliferation, survival and function of mononuclear phagocytes, dendritic cells and osteoclasts. While IL-34 and CSF-1 may induce similar cellular responses, they are differentially expressed and as such exert complimentary actions via CD115.

The AFS98 antibody may be used for identification of myeloid lineage cells by flow cytometry, and is commonly used for in vivo or in vitro neutralization of CSF-1 Receptor (use format appropriate for functional assays).

Type: Primary
Antigen: CSF1R
Clonality: 0
Clone: AFS98
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty