Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mouse IgG2a Monoclonal Isotype Control [clone: C1.18.4] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)

Supplier: Tonbo Biosciences
65-4724-U100
10141-206EA 124 USD InStock
10141-206
Mouse IgG2a Monoclonal Isotype Control [clone: C1.18.4] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)
Antibodies
The C1.18.4 immunoglobulin is useful as an isotype-matched control. The C.1.18.4 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2a antibodies.

Type: Control
Antigen: C1.18.4
Clonality: Monoclonal
Clone: C1.18.4
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: 0

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty