Would you like to log in again?
VISION 10™ Polyester Knit Cleanroom Wipers, High-Tech Conversions
Supplier: High-Tech Conversions
VS10-99 VS10-98
10141-364BG 58.34 USD InStock
10141-364 10141-362
VISION 10™ Polyester Knit Cleanroom Wipers, High-Tech Conversions
Wipes Cleanroom Wipes Dry Cleanroom Wipes
VISION 10™ wipers are extremely low in particle generation and chemical extractables. The fabric consists of premium quality virgin polyester fibers that are continuously filamented throughout the knit construction.
  • Lint-free
  • Cleanroom class 100 (ISO Class 5)
  • Cleanroom laundered
  • Polyester
  • Individually inspected
  • Abrasion resistant
  • Absorbant
VISION 10™ wipers are knitted on state-of-the-art equipment especially for use in cleanrooms. This fabric undergoes a multitude of special processes that include cleaning, scouring, cutting and a special proprietary cleaning process.
Most importantly, VISION 10™ wipers are laundered in a class 100 cleanroom using ultra-filtered water of 0.45 µ purity and HEPA-filtered air during moisture removal.
All VISION 10™ wipers are individually inspected with a lot-to-lot traceability and using ISO 9000 standards.
Ordering information: Wipers are packaged either 150 or 500 per bag, double-bagged, with 10 bags per case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty