Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Unisex Technology Lab Coat, Reusable
Supplier: VWR
VWR®
10141-374 10141-346 10141-344 75819-554 75819-556 75819-564 75819-566 75819-574 75819-560 75819-558 10141-372 10141-342 10141-350 10141-370 10141-348 75819-562
75819-560CS 1096.84 USD InStock
75819-560 75819-574 75819-562 75819-558 10141-370 75819-554 75819-564 75819-556 75819-566 10141-346 10141-344 10141-374 10141-342 10141-372 10141-350 10141-348
VWR® Unisex Technology Lab Coat, Reusable
Workwear
Unisex Technology lab coats are designed for use in research environments, quality-control laboratories, and industrial areas.
  • Plain front with no embroidery
  • Industrial and washable
  • Durable
  • Comfortable
  • Poly/cotton fabric (80/20 blend)
  • Available in white and navy blue
The coats feature knit cuffs and a headphone port in the pocket.
Additionally, the coats have side vents for easy access to pant pockets. The overall length is 99 cm (39").
Lab coats are manufactured from durable and comfortable poly/cotton fabric (80/20 blend) for long-term wear. These industrial, washable garments feature a quick-release five-snap front, one breast pocket with a pencil slot, and two oversized hip pockets large enough for a mini-tablet.
The navy blue coats feature a lapel collar with an additional snap on the collar.
Ordering information: Sizes 4X Large and 5X Large are also available for special order. Contact your Avantor sales representative for more information.
Delivery information: Please allow up to four weeks for delivery.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty