Would you like to log in again?
8 TUBE HOLDER QTY 1 PP
Supplier: QSONICA, LLC.
Holder^ Holder and stem 8 tube, accessory for the 431C2 Cup Horn. holder accomodates 1. 5ml polypropylene sample tubes.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
8 TUBE HOLDER QTY 1 PP 10141-476 451 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty