Would you like to log in again?
Septa, Thermo Scientific
Supplier: Thermo Scientific
31303250-BP 313P3240 313P3211 31303230 31303215-BP 313P3215-BP 313G3230 313B3240 313G3228 313P3233 31303240 31303233-BP 313G3215-BP 313P3233-BP 313B3233 313P3240-BP 31303211 313G3240-BP 31303230-BP 313G3230-BP 313G3211 31303228 313G3240 31303240-BP
10141-480PK 134.09 USD InStock
10141-480 10056-086 10056-084 10141-490 10056-068 10056-088 10056-066 10057-432 10141-488 10057-442 10057-434 10141-486 10141-498 10141-484 10056-070 10141-496 10141-482 10057-438 10056-080 10141-494 10056-090 10141-504 10057-440 10141-502
Septa, Thermo Scientific
Septa
Thermo Scientific™ BTO and TR-Green septa for GC instruments.
  • Variety of materials and temperature limits for all applications
  • For use in Thermo Scientific™ TRACE™ and FOCUS™ GCs and instruments from other manufacturers
  • Contaminant-free in individual blister packs
Made of low-bleed silicone, these septa have excellent mechanical properties, are ideal for demanding GC-MS applications, and may be used reliably up to 400 °C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty