Would you like to log in again?
Lab-Sorb™ for Liquid Disposal, Diversified Biotech
Supplier: Diversified Biotech
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SORB-2100 SORB-3100 SORB-2200 SORB-2000 SORB-3200 SORB-2300 SORB-1000 SORB-3000
10141-808KT 57.25 USD InStock
10141-808 10141-806 10141-818 10141-816 10141-814 10141-812 10141-820 10141-810
Lab-Sorb™ for Liquid Disposal, Diversified Biotech
Spill Control Materials
  • Converts liquid waste to solid waste in seconds
  • Lab-Sorb granules can rapidly absorb aqueous or mixed aqueous and organic solvent waste
  • One 16 oz. jar can absorb up to 50 liters of liquid
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty