Would you like to log in again?
TubePopper, Diversified Biotech
Supplier: Diversified Biotech
TPOP-1000
10141-898EA 23.06 USD InStock
10141-898
TubePopper, Diversified Biotech
Crimpers, Cappers and Decappers
For one-handed opening and closing of microtubes
  • “Pry open” design means tubes pop right open, saving your fingers from the repetitive stress of opening microtubes
  • Bottom portion of TubePopper allows caps to be pressed closed without snagging glove fingers
Works with most flat-top and domed microtubes. TubePopper can be worn on any finger so you can find the perfect pipetting fit.
Ring design allows researcher to wear the TubePopper on the pipetting hand.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty